Learn More
BOLA Gaskets for Caps
Product Code. 17147434
Change view
Click to view available options
Product Type:
Septa
Unit Size:
Pack of 5
Product Code. Product Type Unit Size
17147434 Septa Pack of 5
1 options
Product Code. 17147434 Supplier BOLA Supplier No. H97345
  • ONLINE EXCLUSIVE PRICE
    16.80 €
    12.08 € / Pack of 5
    Save 4.72 €
    28% Off
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Designed specifically for use with BOLA screw caps with apertures, serving as seal or septum replacements
  • Upper side made of PTFE, lower side made of silicone-elastomer for balancing unevennesses on sealing surfaces
  • After assembly, the product is only exposed to PTFE
  • Abmessungen: For thread GL 45 Dia. of gasket 43.2 mm, Thickness of gasket 3.3 mm
Specifications
Color White
Material PTFE-faced Silicone
Sterility Non-sterile
Autoclavable Not Autoclavable
Shape Round
Product Type Septa