Learn More
BOLA Gaskets for Caps
Product Code. 31891578
Change view
Click to view available options
Product Type:
Septa
Unit Size:
Pack of 20
Product Code. Product Type Unit Size
31891578 Septa Pack of 20
1 options
Product Code. 31891578 Supplier BOLA Supplier No. H97345
  • ONLINE EXCLUSIVE PRICE
    61.63 €
    49.60 € / Pack of 20
    Save 12.03 €
    19% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed specifically for use with BOLA screw caps with apertures, serving as seal or septum replacements
  • Upper side made of PTFE, lower side made of silicone-elastomer for balancing unevennesses on sealing surfaces
  • After assembly, the product is only exposed to PTFE
  • Abmessungen: For thread GL 45 Dia. of gasket 43.2 mm, Thickness of gasket 3.3 mm
Specifications
Color White
Material PTFE-faced Silicone
Sterility Non-sterile
Autoclavable Not Autoclavable
Shape Round
Product Type Septa