missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrodin
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 62499-27-8 |
| Melting Point | 169.0°C to 170.0°C |
| Molecular Formula | C13H18O7 |
| Quantity | 1 g |
| InChI Key | PUQSUZTXKPLAPR-UJPOAAIJSA-N |
| SMILES | OC[C@H]1O[C@@H](OC2=CC=C(CO)C=C2)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[4-(hydroxymethyl)phenoxy]oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 286.28 |
| Formula Weight | 286.28 |
| Chemical Name or Material | Gastrodin |
Safety and Handling
Product Identifier
- Gastrodin
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 4-(Hydroxymethyl)phenyl .beta.-D-glucopyranoside
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction