missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Gatifloxacin

Product Code. 17110122
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17110122 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17110122 Supplier STEMCELL Technologies Supplier No. 72752
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibiotic; Inhibits bacterial DNA gyrase and topoisomerase IV

  • Gatifloxacin is a fluoroquinolone antibiotic which inhibits bacterial DNA gyrase (IC50 = 0.109 μg/ml) and topoisomerase IV (IC50 = 13.8 μg/ml; Takei et al.) It is much less effective against HeLa cell topoisomerase II (IC50 = 265 μg/ml; Takei et al.)
  • Promotes self-renewal in cultured human and mouse embryonic stem cells (Desbordes et al.)

Specifications

CAS 112811-59-3
Synonym AM 1155; BMS 206584-01; PD 135432
Quantity 1 g
Molecular Formula C19H22FN3O4
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.