missing translation for 'onlineSavingsMsg'
Learn More

Gatifloxacin, 98%, Thermo Scientific Chemicals

Product Code. 15549513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15549513 25 g 25g
15529513 1 g 1g
15539513 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15549513 Supplier Thermo Scientific Acros Supplier No. 460230250
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112811-59-3
Molecular Formula C19H22FN3O4
Molecular Weight (g/mol) 375.40
MDL Number MFCD00895399
InChI Key XUBOMFCQGDBHNK-UHFFFAOYNA-N
Synonym gatifloxacin, tequin, zymar, gatiflo, gatifloxacine, gatiquin, gatispan, gatilox, zymaxid, gaity
PubChem CID 5379
ChEBI CHEBI:5280
IUPAC Name 1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
SMILES COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1

Specifications

Melting Point 185.0°C to 187.0°C
Loss on Drying 4% to 8%
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 375.39
Percent Purity 98%
Chemical Name or Material Gatifloxacin
compliance-icons
Product Identifier
  • Gatifloxacin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.