missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ BP624 GC Capillary Columns

SGE BP624 GC Capillary Columns - A polar phase designed for EPA volatile organics analysis

Brand:  Trajan™ 054836

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12760720

  • 850.00 € / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

  • Polar phase
  • EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
  • Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications

Specifications

0.53 mm
230/240°C
3 mm
High polar
BP624 GC Capillary Column
0°C to 240°C
30 mm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers