Learn More
Trajan™ BP624 GC Capillary Columns
SGE BP624 GC Capillary Columns - A polar phase designed for EPA volatile organics analysis
Brand: Trajan™ 054832
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Polar phase
- EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
- Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
0.32 mm | |
230/240°C | |
1.8 μm | |
30 m |