missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GeA-69

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GeA-69 is a selective allosteric inhibitor of poly (ADP-ribose) polymerase 14 (PARP14) macrodomain 2 (MD2) (IC50 = 720 nM, Kd = 860 nM). It inhibits adenosine diphosphate ribose (ADPR) binding to the macrodomain 2 of PARP14 and prevents PARP14 MD2 localization to DNA damage sites in intact cells. GeA-69 is highly cell permeable.
Specifications
Specifications
| Chemical Name or Material | N-[2-(9 H-Carbazol-1-yl)phenyl]acetamide |
| CAS | 2143475-98-1 |
| Quantity | 10 mg |
| Target | PARP |
| Molecular Formula | C20H16N2O |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction