missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GeA-69
GREENER_CHOICE

Product Code. 18718866
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18718866

Brand: Tocris Bioscience™ 6795/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective allosteric PARP14 inhibitor

GeA-69 is a selective allosteric inhibitor of poly (ADP-ribose) polymerase 14 (PARP14) macrodomain 2 (MD2) (IC50 = 720 nM, Kd = 860 nM). It inhibits adenosine diphosphate ribose (ADPR) binding to the macrodomain 2 of PARP14 and prevents PARP14 MD2 localization to DNA damage sites in intact cells. GeA-69 is highly cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-(9 H-Carbazol-1-yl)phenyl]acetamide
CAS 2143475-98-1
Quantity 50 mg
Target PARP
Molecular Formula C20H16N2O
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.