Learn More
Biodesign™ Gel storage system GelFile binder and 20 gel protector sheets 2 gels per sheet for gels up to 180mm x 255mm BioDesign
Product Code. 12896502
Change view
Click to view available options
Gel Size:
18 x 25.5 cm
Unit Size:
Each
Product Code. Gel Size unitSize
12896502 18 x 25.5 cm Each
1 options
Product Code. 12896502 Supplier Biodesign™ Supplier No. G303
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Stores, organises and displays dried polyacrylamide and agarose gels
  • Each BioDesign GelFile™ binder contains heavy crystal clear vinyl pages with pockets and white backgrounds
  • The pages are chemically neutral and are guaranteed not to alter your dried gels or turn yellow for TEN years
  • Dried gels are held flat for long term storage without curling
  • Shields gels from moisture, accidental spills and possible damage
  • White background allows optimal comparison between gels
  • Each BioDesign GelFile™ binder holds 20 to 25 pages
  • A clear label holder on the spine allows you to personalise each binder
Tekniske data
Gel Size 18 x 25.5 cm
Quantity 1 Binder, 20 Gel Protector Sheets
Product Type Gel Storage System, Gel Storage Binder
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.