missing translation for 'onlineSavingsMsg'
Learn More

Gelatin, for analysis, granular

Product Code. 10075660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10075660 500 g Plastic bottle 500g
10453655 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10075660 Supplier Thermo Scientific Acros Supplier No. 410875000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Gelatin, CAS # 9000-70-8, also known as gelatin type A, derived from porcine skin, is a gelling compound used in electrophoresis and in vitro cell cultures. | CAS: 9000-70-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Gelatin derived from porcine skin is a powder created from the acidic digestion of collagen and is also known as gelatin type A

• It comprises heterogeneous mixture of water-soluble proteins of high average molecular masses, including glycine, proline and hydroxyproline

Applications

• Gelatin has been used in in vitro coating cell culture to improve attachment of cell lines • It is added to PCR reactions in order to stabilize DNA Taq polymerase

• It can be used as a blocking reagent in several biological techniques including western blotting, ELISA, and immunochemistry

• It can be applied in microencapsulation assays to delivery vehicle for release of active biomolecules since it is a biocompatible polymer

• It is used as a component of media for species differentiation in bacteriology and in light microscope autoradiography

• It has been used in the generation of scaffolds for tissue engineering assays

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638

Specifications

Color Beige to Yellow
pH 3.8 to 7.6 (1% soln., about 55°C) (PHEUR)
Loss on Drying 13% max. (105°C)
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 500 g
Merck Index 15, 4415
Grade Analysis
Physical Form Crystalline Powder
Chemical Name or Material Gelatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.