missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GelBond PAG Gel Film

Product Code. 10754037 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Application):
Agarose gels
Polyacrylamide gels
Length (Metric):
94 mm
258 mm
260 mm
Width (Metric):
84 mm
124 mm
203 mm
Unit Size:
Pack of 50
Product Code. For Use With (Application) Length (Metric) Width (Metric) unitSize
10754037 Polyacrylamide gels 260 mm 203 mm Pack of 50
10086234 Polyacrylamide gels 258 mm 124 mm Pack of 50
10075804 Agarose gels 258 mm 124 mm Pack of 50
10575785 Agarose gels 94 mm 84 mm Pack of 50
4 options
This item is not returnable. View return policy

Product Code. 10754037

Brand: Cytiva 80112937

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cytiva™ GelBond PAG Gel Film is a high density homogenous medium.

Cytiva™ GelBond Film is a transparent, flexible polyester film designed to support gels. Gels cast on GelBond Film remain permanently attached to the film through electrophoresis or immunodiffusion and all subsequent fixing, staining, destaining, and drying procedures.
  • GelBond Film is made from thin polyester (0.2mm). Pack of 50 films, 124 x 258mm.
  • High-density homogenous medium
Each film has one hydrophilic side and one hydrophobic side. The gel must be cast on the hydrophilic side, which is the side where a water droplet will spread out.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Polyacrylamide gels
Quantity 50 Pack
Length (English) 10.2 in.
Length (Metric) 260 mm
Width (English) 7.9 in.
Width (Metric) 203 mm
Product Type Gel Film
compliance-icons
Product Identifier
  • Blue Sepharose High Performance, 50 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.