missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Geldanamycin

Product Code. 17140312
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17140312 1 mg 1mg
17150312 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17140312 Supplier STEMCELL Technologies Supplier No. 74032
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inhibits Hsp90

  • Geldanamycin is a benzoquinone ansamycin antibiotic that selectively inhibits heat shock protein 90 (Hsp90) (Schulte et al.; Whitesell et al.) It inhibits the ATPase activity of Hsp90 by binding with high affinity to the N-terminal ATP-binding site (Kd = 1.2 μM; Roe et al.) Geldanamycin also binds to the glucose-regulated protein GRP94, an endoplasmic reticulum protein with homology to Hsp90
  • This interaction destabilizes its complex with p185-ERBB2 leading to degradation of ERBB2 protein, which is overexpressed in aggressive breast cancers (Chavany et al.; Castagnola et al.)
  • Inhibits growth of 60 human tumor cell lines (Supko et al.)
  • Regulates innate immune response as demonstrated by inhibition of inflammasome activity in a mouse gout model (Mayor et al.)
  • Inhibits replication of HSV-1 and HSV-2 (Li et al.)

Specifications

CAS 30562-34-6
Synonym NSC 122750
Quantity 1 mg
Molecular Formula C29H40N2O9
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.