missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Geldanamycin
Description
- Geldanamycin is a benzoquinone ansamycin antibiotic that selectively inhibits heat shock protein 90 (Hsp90) (Schulte et al.; Whitesell et al.) It inhibits the ATPase activity of Hsp90 by binding with high affinity to the N-terminal ATP-binding site (Kd = 1.2 μM; Roe et al.) Geldanamycin also binds to the glucose-regulated protein GRP94, an endoplasmic reticulum protein with homology to Hsp90
- This interaction destabilizes its complex with p185-ERBB2 leading to degradation of ERBB2 protein, which is overexpressed in aggressive breast cancers (Chavany et al.; Castagnola et al.)
- Inhibits growth of 60 human tumor cell lines (Supko et al.)
- Regulates innate immune response as demonstrated by inhibition of inflammasome activity in a mouse gout model (Mayor et al.)
- Inhibits replication of HSV-1 and HSV-2 (Li et al.)
Specifications
Specifications
| CAS | 30562-34-6 |
| Synonym | NSC 122750 |
| Quantity | 5 mg |
| Molecular Formula | C29H40N2O9 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?