missing translation for 'onlineSavingsMsg'
Learn More

Gellan Gum Powder, MP Biomedicals™

Product Code. 11425853 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Product Code. Quantity unitSize
11425853 100 g 100g
11435853 500 g 500g
11400666 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11425853

Brand: MP Biomedicals 02180106.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a gelling agent in place of agar in microbiological media. Gellan Gum Powder, MP Biomedicals is a water-soluble polysaccharide produced by Pseudomonas elodea, a bacterium. It is used in a variety of immobilization matricies and as a food additive,

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71010-52-1
Molecular Formula (C24H38O20)A
Synonym Agar substitute gelling agent

Specifications

Quantity 100 g
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.