missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix

Product Code. 12053569
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12053569 5 mL 5mL
11692139 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12053569 Supplier Gibco™ Supplier No. A1413202
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

A soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. It is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation.

Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors and is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation. The major components of Geltrex™ matrix include laminin, collagen IV, entactin, and heparin sulfate proteoglycans. It is ideal for the promotion and maintenance of differentiated phenotypes in a variety of cell cultures including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC). Geltrex™ matrix offers, on average, a 2-fold reduction in several growth factors. As a result, analysis of specific growth factor effects in applications requiring basement membrane extract can be carried out in a controlled and reproducible format.

Key Features of Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix:

  • Free of lactose dehydrogenase elevating virus (LDEV) making it ideal for all types of cell culture and mouse in vivo applications
  • Consistent protein concentration lot-to-lot helps reduce the need to screen lots
  • Protocols for various applications, including tube formation assays, are available

Gain Reliability and Consistency in Your hESC Studies

Recent advances in processing EHS tumor cells have resulted in virus free basement membrane extracts. Every lot is tested by RT-PCR methods so you can now have confidence that every lot of Geltrex™ is virus free. Additionally, our consistent manufacturing process helps ensure a protein concentration with very little variation from lot-to-lot, averaging 15 mg/ml so you can have confidence in every purchase.

Investigate Angiogenesis with Validated Gibco™ Protocols

There are many applications for Geltrex™ LDEV-Free reduced growth factor basement membrane matrix, each requiring different thicknesses and concentrations. In general, a protein concentration of >5 mg/ml is used for differentiation studies of primary cells. Geltrex™ LDEV-Free reduced growth factor basement membrane matrix has been used to develop tube formation assay protocols using Gibco™ primary cells and specialty media.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Variety of cultured cells, including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC)
Classification Reduced Growth Factor
Concentration 12 to 18 mg/mL
Form Frozen
Product Type Basement Membrane Matrix
Serum Level Serum-free
Sterility Sterile
Without Additives No Phenol Red
Product Line Gibco
Quantity 5 mL
Shipping Condition Dry Ice
Endotoxin Level Low
Format Tube(s)
Purity or Quality Grade Cell Culture Grade
Species Mouse
Content And Storage Store at -20°C.
Culture Type 3D Cell Culture, Mammalian Cell Culture, Neuronal Cell Culture, Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic), Adherent Cell Culture
Show More Show Less
What growth factors are reduced in the Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413202)?

Although the standard basement membrane extract (BME) contains growth factors such as EGF, IGF‐1, PDGF and TGF‐beta, the reduced growth factor basement membrane extract product (Geltrex matrix) contains below detectable levels of EGF, PDGF and NGF. It still has some TGF‐beta (about 2 ng/mL) and 3 times less IGF‐1 on average than regular BME, hence the designation "reduced". Neither standard nor reduced BME contain NGF or FGF‐2.

What volume do you recommend for thin coating 35 mm dishes with Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413201, A1413202) or Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301, A1413302)?

We recommend thin (non-gelling) coating 35 mm dishes with 1.5 mL of Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413201, A1413202) or Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301, A1413302) prediluted 1/100 in DMEM/F-12 medium (1% final concentration).

Can I refreeze and reuse Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix after having used it in an experimental setup?

Yes, you can refreeze and reuse Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix. However, you should avoid multiple freeze/thaw cycles. hence, we would recommend the following:

  1. Thaw Geltrex Matrix in a refrigerator overnight at 2-8 degrees C.
  2. Mix Geltrex Matrix solution by slowly pipetting up and down. Be careful not to introduce air bubbles.
  3. If not using up the complete solution, prepare multiple aliquots, either of the original solution or the diluted one, and freeze.

How do I characterize human embryonic stem ( ES) cells?

Human ES cells are generally characterized by their typical morphology (they grow as tightly packed clusters of small cells with high ratio of nucleus to cytoplasm); surface marker expression; RT-PCR detection of stem cell-specific gene expression (such as Oct3/4, Sox2, and Nanog); alkaline phosphatase staining, and telomerase activity assay. The most commonly used ES specific surface markers include stage-specific embryonic antigens SSEA-3 and SSEA-4 for human ES cells. Other ES-specific surface antigens also include TRA-1-60 and TRA-1-81. (Science 282:1145 (1998).

How are human embryonic stem (ES) cells derived?

Human ES cells are derived from human blastocyst inner cell masses, isolated by immunosurgery with rabbit antiserum to BeWO cells (a human trophoblast cell line) (Science 282:1145 (1998)).

What are ES cells?

Embryonic stem (ES) cells are derived from the early mammalian embryo and are capable of unlimited, undifferentiated proliferation in vitro while maintaining their potential to differentiate into a wide of range of adult tissues including germ cells. The pluripotency of the ES cells is normally demonstrated in vitro by inducing ES cells to differentiate into embryoid bodies and checking lineage-specific markers for differentiated cells in three body layers (endo, meso, and ectoderm), or injecting them into immunodeficient mice and determining the cell types produced in the teratomas.

Can differentiation of cells occur when using Gibco Geltrex Matrix? Can this be prevented?

Yes, try a variety of dilutions to coat the plates. Some customers use a 1:100 dilution and have better results. The optimal dilution is cell-line dependent.

Why can Gibco Geltrex Matrix be used for adhesion-related research, as opposed to Gibco AlgiMatrix 3D Culture System which cannot?

Geltrex Matrix is a basement membrane extract that contains collagen IV, laminin, and other matrix proteins and growth factors that are required for cell adhesion. AlgiMatrix Matrix contains only alginate which does not facilitate cell adhesion. There is nothing in the alginate sponges for the cells to interact with.

Is it normal to see floating debris when pre-coating with Ready-to-Use Gibco Geltrex Matrix?

Yes, this is normal. It has been observed in product development. Remove the Gibco Geltrex Matrix solution after pre-coating and add cells as normal. H9 human embryonic stem cells have been cultured on dishes pre-coated with Ready-to-Use Gibco Geltrex Matrix with this observation in the pre-coating procedure.

What is LDEV?

Lactate dehydrogenase-elevating virus (LDEV) infects mouse macrophages and induces persistent infections in laboratory mice. LDEV, an RNA virus, alters in vivo physiology, and is a common contaminant of biological materials derived from transplantable tumor cell lines.

Can cells be removed from the Gibco Geltrex Matrix?

If you would like to culture the cells once recovered from the matrix, we recommend washing the cells first, then using trypsin/EDTA, TrypLE reagent, or a solution of 3,000 U/mL collagenase and 1,000 U hyaluronidase in low-glucose DMEM. If you would like to monitor cell surface markers, we recommend removal of the cells by mechanical means, such as a vigorous wash or a delicate scrape.

Note that the phenotype of the cells may change if they are cultured in 2D after their removal from the matrix.

Has Gibco Geltrex Matrix been tested with StemPRO hESC serum-free Medium?

Yes, these products were designed to work together.

What is Gibco Geltrex Matrix?

Gibco Geltrex Matrix is a soluble form of reduced growth factor (RGF) basement membrane extract (BME) purified from murine Engelbreth-Holm-Swarm tumor. Historically these preparations have contained the lactate dehydrogenase-elevating virus (LDEV), which does not impact the performance of the material but may be of concern in certain applications such as xenografts or other clinically focused research. New manufacturing methods are now available that allow for the removal of this virus. The resulting LDEV-free material is tested using PCR methods to confirm that the material is virus-free.

What are some general differences between Gibco Geltrex Matrix, collagen, AlgiMatrix 3D Culture System, and CTS Cellstart Substrate?

Gibco Geltrex Matrix and collagen (rat tail and bovine) can be used as either a coating solution or a 3D gel matrix. CELLstart Substrate was developed to be used as a xeno-free coating matrix for only ESC applications (as a substitute for Gibco Geltrex Matrix or Matrigel Matrix, which are of animal origin). AlgiMatrix Matrix is a 3D scaffold-type matrix that does not support cell attachment but does provide a good environment for growing spheroids that can be easily harvested for downstream applications.


compliance-icons
Product Identifier
  • 2-Bromo-5-chlorotoluene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromo-5-chlorotoluene

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.