missing translation for 'onlineSavingsMsg'
Learn More

Gemcitabine elaidate, MedChemExpress

Product Code. 16438427
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438427 5 mg 5mg
16421907 50 mg 50mg
16415748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16438427 Supplier Medchem Express Supplier No. HY13538
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.

Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.

Specifications

Chemical Name or Material Gemcitabine elaidate
CAS 210829-30-4
Color White
Purity Grade Notes Research
Molecular Formula C27H43F2N3O5
Quantity 5 mg
Synonym CP-4126 CO-101 Gemcitabine 5'-elaidate
Solubility Information DMSO : ≥ 26 mg/mL (49.28 mM)
SMILES O[C@@H](C(F)(F)[C@H](N1C(N=C(C=C1)N)=O)O2)[C@H]2COC(CCCCCCC/C=C/CCCCCCCC)=O
Molecular Weight (g/mol) 527.64
Formula Weight 527.64
Percent Purity 97.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.