missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Gemcitabine hydrochloride

Product Code. 16491986
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491986 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491986 Supplier MedChemExpress Supplier No. HYB0003
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Gemcitabine Hydrochloride (LY 188011 Hydrochloride) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. Gemcitabine Hydrochloride inhibits DNA synthesis and repair, resulting in autophagyand apoptosis.

Specifications

Molecular Weight (g/mol) 299.66 g/mol
Product Type Reference compound
Content And Storage 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Shipping Condition Room temperature
Purity 98.0%
Quantity 500 mg
Solubility Information H2O : ≥ 66.66 mg/mL (222.45 mM) ∣DMSO : 25 mg/mL (83.43 mM∣ultrasonic and warming and heat to 60°C) ∣DMF : 2.5 mg/mL (8.34 mM∣Need ultrasonic)
For Use With (Application) Cancer-programmed cell death
CAS 122111-03-9
Synonym LY 188011 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.