missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Gemcitabine hydrochloride
GREENER_CHOICE

Product Code. 30001393
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001393 10 mg 10mg
30001403 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001393 Supplier Tocris Bioscience™ Supplier No. 3259/10
  • ONLINE EXCLUSIVE PRICE
    216.00 €
    203.70 € / 10mg
    Save 12.30 €
    5% Off
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

DNA synthesis inhibitor

Deoxycytidine analog that inhibits DNA synthesis. Metabolized to form gemcitabine triphosphate (dFdCTP) and gemcitabine diphosphate (dFdCDP). dFdCTD inhibits ribonucleotide reductase causing a reduction in cellular nucleotides. dFdCTP is incorporated in DNA resulting in DNA strand termination. Displays antitumor activity in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122111-03-9
Molecular Formula C9H12ClF2N3O4

Specifications

Quantity 10 mg
Formula Weight 299.66
Purity >99%
Molecular Weight (g/mol) 299.659
Chemical Name or Material Gemcitabine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.