missing translation for 'onlineSavingsMsg'
Learn More

Gemcitabine, TRC

Product Code. 18053730
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18053730 1 g 1g
18051330 100 mg 100mg
17949729 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18053730 Supplier Toronto Research Chemicals Supplier No. TRCG3050281G
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95058-81-4
Molecular Formula C9H11F2N3O4
Molecular Weight (g/mol) 263.2
Synonym 2'-Deoxy-2',2'-difluorocytidine, 2',2'-Difluoro-2'-deoxycytidine, 2',2'-Difluorodeoxycytidine, DDFC, DFdC, DFdCyd, Folfugem, GemLip, Gemcel, Gamcitabine, LY 188011, NSC 613327, Zefei;
IUPAC Name 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)C2(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H11F2N3O4/c10-9(11)6(16)4(3-15)18-7(9)14-2-1-5(12)13-8(14)17/h1-2,4,6-7,15-16H,3H2,(H2,12,13,17)/t4-,6-,7-/m1/s1
Formula Weight 263.0718
Chemical Name or Material Gemcitabine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.