missing translation for 'onlineSavingsMsg'
Learn More

Gemfibrozil, TRC

Product Code. 30138578
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138578 50 mg 50mg
30136064 1 g 1g
30007707 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138578 Supplier Toronto Research Chemicals Supplier No. TRCG30575050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25812-30-0
Molecular Formula C15 H22 O3
Molecular Weight (g/mol) 250.33
Synonym Gemfibrozil, 2,2-Dimethyl-5-(2,5-xylyloxy)valeric acid, 5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid, CI 719, Decrelip, Genlip, Gevilon, Lipur, Lopid, Lopizid, Trialmin 900
IUPAC Name 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
SMILES Cc1ccc(C)c(OCCCC(C)(C)C(=O)O)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H22O3/c1-11-6-7-12(2)13(10-11)18-9-5-8-15(3,4)14(16)17/h6-7,10H,5,8-9H2,1-4H3,(H,16,17)
Formula Weight 250.1569
Chemical Name or Material Gemfibrozil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.