missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Buffer

Product Code. 10125605 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1.5 mL
6 x 1.5 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10125605 6 x 1.5 mL Each
10219244 1.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10125605 Supplier Applied Biosystems™ Supplier No. N8080129
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applied Biosystems GeneAmp 10X PCR Buffer I is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification.

Applied Biosystems™ GeneAmp™ 10X PCR Buffer I is optimized for use with AmpliTaq™ DNA Polymerase, resulting in robust PCR amplification.

  • Optimized for use with AmpliTaq™ DNA Polymerase, offering maximum PCR performance
  • Available in several convenient sizes

GeneAmp™ 10X PCR Buffer I contains 15 mM MgCl2. When diluted 1:10, it provides the optimal pH and ionic strength for PCR amplification with AmpliTaq™ DNA Polymerase.

Note: See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PCR Buffer
Concentration 10X
For Use With (Application) Standard PCR
Quantity 6 x 1.5 mL
Includes • GeneAmp 10X PCR Buffer I (with 15 mM MgCl2), 6 x 1.5 mL

Store at -15°C to -25°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.