missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GeneArt™ High-Order Vector Conversion Cassette

Product Code. 11350952
Change view
Click to view available options
Quantity:
10 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11350952 10 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11350952 Supplier Invitrogen™ Supplier No. A13291

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A ready-to-use, 3.8kb linear BAC/YAC cassette, containing yeast replication and selection elements that can make any E.coli vector compatible with the GeneArt™ High-Order Genetic Assembly System

The GeneArt™ High-Order Vector Conversion Cassette is a ready-to-use, 3.8 Kb linear BAC⁄YAC cassette, containing yeast replication and selection elements that can make any E.coli vector compatible with the GeneArt™ High-Order Genetic Assembly System (cat# A13285 & A13286). Creating your compatible vector is as simple as blunt cloning the cassette into the vector’s multiple cloning site. This product only includes the DNA cassette.

Some of the key features and elements of the Conversion Cassette include:
  • TRP1 to allow selection of yeast transformants in medium lacking tryptophan
  • URA3 as an optional counter selectable marker in yeast
  • ARS⁄CEN origin to allow stable maintenance of the construct in yeast
  • Chloramphenicol resistance for selection in E.coli
  • 6 restriction enzyme sites, including 2 endonuclease homing sites, designed for rare-cutters to simplify the linearization of the final construct
If you prefer to use a commercial ready-to-use vector, please consider the GeneArt™ pYES1L Vector with Sapphire Technology™ (cat# A13287). The vector comes linearized and has been verified to clone up to 110 Kb of DNA fragments.

For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Resistance Bacterial Chloramphenicol (CmR)
Cloning Method Seamless Cloning
Content And Storage The product contains one vial of linear conversion cassette at a concentration of 100 ng⁄ml. Each vial contains enough DNA to convert 10 different E.coli vectors.

Store the product at -20°C.

The cassette is guaranteed stable for 6 months when properly stored.
For Use With (Application) Seamless Cloning & Genetic Assembly
No. of Reactions 10
Product Line GeneArt
Product Type High-Order Vector Conversion Cassette
Quantity 10 Reactions
Replication Origin ARS⁄CEN
Selection Marker Eukaryotic URA3, TRP1
Shipping Condition Dry Ice
Vector pYES1L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.