missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ CALCRL:RAMP1-CRE-bla FreeStyle™ 293F Cells

Product Code. 10164363
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10164363

Brand: Thermo Scientific™ K1437

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ CALCRL:RAMP1-CRE-bla FreeStyle™ 293F cells contain the human Calcitonin Gene Related Peptide Receptor (CALCRL:RAMP1), stably integrated into the CellSensor™ CRE-bla Freestyle™ 293F cell line.

GeneBLAzer™ CALCRL:RAMP1-CRE-bla FreeStyle™ 293F cells contain the human Calcitonin Gene Related Peptide Receptor (CALCRL:RAMP1), (Accession No. NM_005795.3 NM_005855.2) stably integrated into the CellSensor™ CRE-bla Freestyle™ 293F cell line. CellSensor™ CRE-bla Freestyle™ 293F cells (Cat. No. K1130) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element (CRE).

CALCRL:RAMP1-CRE-bla FreeStyle™ 293F cells are functionally validated for Z'-factor and EC50 concentrations of Alpha-CGRP. In addition, CALCRL:RAMP1-CRE-bla FreeStyle™ 293F cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_005855.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ CALCRL:RAMP1-CRE-bla Freestyle™ 293F cells
• GeneBLAzer™ CALCRL:RAMP1-CRE-bla Freestyle™ 293F Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ CALCRL:RAMP1-CRE-bla Freestyle™ 293F cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Description GeneBLAzer™ CALCRL:RAMP1-CRE-bla FreeStyle™ 293F Cells, 8 x 106 Cells, ∽8,000,000 cells/vial at 8,000,000 cells/mL Quantity of Cells, FreeStyle™ 293-F Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
Gene Symbol CALCRL:RAMP1
Includes GeneBLAzer™ CALCRL:RAMP1-CRE-bla FreeStyle™ 293F Cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ CALCRL:RAMP1-CRE-bla Freestyle™ 293F cells, GeneBLAzer™ CALCRL:RAMP1-CRE-bla Freestyle™ 293F Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line Freestyle, GeneBLAzer
Quantity 8 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Sample Type FreeStyle™ 293-F
Target Entry CALCRL:RAMP1
Type Cell Line
Cell Line FreeStyle 293-F Cells
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.