Learn More
Thermo Scientific™ GeneBLAzer™ Gα15-NFAT-bla CHO-K1 Cells
Description
The GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cell line contains the promiscuous Gα protein, Gα15, stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of an NFAT response element stably integrated into CHO-K1 cells. The Gα15-NFAT-bla CHO-K1 parental cell line is designed for the construction of Gi/o coupled GPCR cell based assays.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Description | GeneBLAzer™ Gα±15-NFAT-bla CHO-K1 Cells, 2 x 106 cells, ∽2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | Pharma and biopharma, target and lead identification and validation, Cell analysis, cellular imaging, cell-based reporter assays, drug discovery and development, target and lead identification and validation, g-protein coupled receptor biology, cell-based GPCR reporter assays |
| Gene Symbol | Gα15 |
| Includes | Promiscuous Gα± protein, Gα±15, stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line |
| Product Line | GeneBLAzer |
| Quantity | 2 x 106 Cells |
| Reporter Gene | BLA (Beta-Lactamase) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.