missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ Gα15-NFAT-bla HEK 293T Cells

Product Code. 10358763
Change view
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10358763 2 x 106 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10358763 Supplier Thermo Scientific™ Supplier No. K1539
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The GeneBLAzer™ Gα15-NFAT-bla HEK 293T cell line contains the Gα protein Gα15 stably integrated into the CellSensor™ NFAT-bla HEK 293T parental cell line.

The GeneBLAzer™ Gα15-NFAT-bla HEK 293T cell line contains the Gα protein Gα15 stably integrated into the CellSensor™ NFAT-bla HEK 293T parental cell line. NFAT-bla HEK 293T cells contain a beta-lactamase reporter gene under control of an NFAT response element stably integrated into HEK 293T cells. The Gα15-NFAT-bla HEK 293T master cell line is designed for the construction of Gi/o-coupled GPCR cell-based assays (Figure 1).

TRUSTED_SUSTAINABILITY

Specifications

Cell State Dividing Cells
Content And Storage Shipping: Dry Ice
Storage: Liquid Nitrogen
Description GeneBLAzer™ Gα±15-NFAT-bla HEK 293T Cells, 2 x 106 cells, ∽2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, 293T (HEK) Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Sto
Detection Method Fluorescence
For Use With (Application) Cellular imaging, pharma and biopharma, Cell analysis, cellular imaging, cell-based reporter assays, cell-based and beta-lactamase assays, drug discovery and development, target and lead identification and validation, g-protein coupled receptor biology
Gene Symbol Gα15
Includes Gα± protein Gα±15 stably integrated into the CellSensor™ NFAT-bla HEK 293T parental cell line
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Sample Type 293T (HEK)
Type Cell Line
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.