missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ GLP1R-CRE-bla CHO-K1 Cells

Product Code. 10742524
Change view
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10742524 8 x 106 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10742524 Supplier Thermo Scientific™ Supplier No. K1783
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells contain the human Glucagon-like Peptide 1 Receptor (GLP1R) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.

The GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells contain the human Glucagon-like Peptide 1 Receptor (GLP1R) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase reporter gene under control of the CRE response element.

The GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_002062.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells
• GeneBLAzer™ GLP1R-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) GPCRs
Gene Symbol GLP1R
Includes GLP1R-CRE-bla CHO-K1 cells, GLP1R-CRE-bla CHO-K1 Assay Protocol, Certificate of Analysis
Population Status Pool of Stable Cells
Product Line GeneBLAzer
Protein Family GPCR (G-Protein Coupled Receptors)
Quantity 8 x 106 Cells
Readout End Point
Reporter Gene BLA (Beta-Lactamase)
Target Entry GLP1R
Type CHO-K1 Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.