Learn More
Thermo Scientific™ GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 Cells
Description
GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 cells contain the human 5-hydroxytryptamine (serotonin) receptor 2B (HTR2B), (Accession No. NM_000867) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells contain a beta-lactamase reporter gene under control of the NFAT Response Element. The GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of 5-HT (Figure 1). In addition, GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Specifications
| Accession Number | NM_000867.2 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 cells • GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Description | GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 Cells, 8 x 106 Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | FRET |
| Gene Symbol | HTR2B |
| Human Protein Class | G-Protein Coupled Receptor (GPCR) |
| Includes | GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).. Includes: GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 cells, GeneBLAzer™ HTR2B-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.