missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ PROKR2-Gqi5-NFAT-bla CHO-K1 Cells

Product Code. 10074004
Click to view available options
Quantity:
8 x 106 cells/vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10074004

Brand: Thermo Scientific™ K1856

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ PROKR2-Gqi5-NFAT-bla CHO-K1 cells contain the human prokineticin receptor 2 (PROKR2), stably integrated into the GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cell line.

GeneBLAzer™ PROKR2-Gqi5-NFAT-bla CHO-K1 cells contain the human prokineticin receptor 2 (PROKR2), (Accession No. NM_144773) stably integrated into the GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cells (Cat. No. K1725) contain a beta-lactamase reporter gene under control of the NFAT Response Element. The GeneBLAzer™ PROKR2-Gqi5-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Prokineticin (EG-VGEF)

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_144773
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage GeneBLAzer™ cell lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) FRET
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Pool of Stable Cells
Product Line GeneBLAzer
Quantity 8 x 106 cells/vial
Readout End Point
Reporter Gene BLA (Beta-Lactamase)
Target Entry PROKR2
Type Dividing Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.