missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ GeneGnome XRQ Benchtop
GREENER_CHOICE

Product Code. 15819942 Shop All Syngene Products
Change view
Click to view available options
Display:
None
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display unitSize
15819942 None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819942 Supplier Syngene™ Supplier No. GGNOMEXRQNPC
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

GeneGnome XRQ Benchtop is a simple chemiluminescence imaging system for Western Blots.

GeneGnome XRQ is a cost effective, high performance chemiluminescence only imaging system. High performance imaging has been proven to achieve comparable sensitivity to film with the added benefit of using advanced software tools to optimise the imaging of faint bands.

  • Peltier cooled, high resolution 16 bit camera (with 65535 grey scales) capable of producing 4 MP images.
  • Camera chip is a leader in the field with a QE of 73% at 425 nm (peak 77%). T
  • F0.95 fixed lens for rapid acquisition times.
  • Maximum viewing area available is 11 x 8 cm.
  • PC required (not included).

GeneSys is an application driven software designed for rapid image capture of biological samples.

  • Simple icon based user interface usable on Win 10, 8 and 7 platforms.
  • Takes the guess work out of imaging, and produces quick and simple publication quality images.
  • All files produced are a unique SGD format, where only raw data is analysed for quantification of samples.
  • Files can also be saved as TIFF, JPEG, BMP formats.
  • User protocols can be saved to review previous images captured, and easily replicate future imaged gels.
  • All captured images can be simply edited with annotations, edit tools, image sharpen and exported to GeneTools.

GeneTools software provides the user with rapid quantification of samples and goes from blots to analysed data in a few quick steps. Analysis sample types include:

  • Molecular weight
  • Colony counting
  • Spot blot
  • Band matching.

All samples can be quantified, including multiplexed protein fluorescence analysis of up to 5 samples at once. Raw data files can be quickly quantified to produce true data for publication data. High quality reports can be exported in Word and Excel files for simple review.

Both GeneSys and GeneTools software meet the requirements for 21 CFR Part 11 and GLP environments.

The system comes with a 3 year warranty (return to factory), unlimited licences of GeneTools analysis software and twice annual free software updates.

Specifications

Display None
Hertz 50 Hz
Voltage 100/240 V
Depth (English) 16.9 in.
Depth (Metric) 43 cm
Height (English) 17.3 in.
Pixel Size 4 MP
Width (English) 14.8 in.
Height (Metric) 17.3 in.
Width (Metric) 37.5 cm
Lens Type F0.95 Fixed
Packaging Quantity 1 Ea.
Size 11 x 8 cm
Dimensions 43 x 37.5 x 44 cm
For Use With (Application) Chemiluminescence Imaging
Product Line Syngene
Color Blue
Product Type Gel Imaging System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.