missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Stabilized and Fresh Whole Blood RNA Kit

Product Code. 15235106 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 preps
Unit Size:
50 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15235106 50 preps 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15235106 Supplier Thermo Scientific™ Supplier No. K0871
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.

The GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The kit includes a DNase l for the enzymatic removal of genomic DNA. RNA can be isolated from whole blood in less than 30 minutes (including cell lysis and DNase l treatment).

Thermo Scientific GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The kit includes a DNase l for the enzymatic removal of genomic DNA. RNA can be isolated from whole blood in less than 30 minutes (including cell lysis and DNase l treatment).

Highlights

  • Versatile—compatible with fresh and frozen blood, buffy coat, as well as blood collected in PAXgene™and Tempus™ Tubes
  • Convenient—specially optimized Lysis Buffer is suitable for RNA stabilization up to 48 h
  • Efficient—up to 4 μg of total RNA from 500 μL of healthy human blood
  • Pure—isolates intact total RNA free of any RT-qPCR inhibitors

Applications

Isolated total RNA can be used in:

  • RT-PCR
  • RT-qPCR
  • Northern Blotting

Includes

  • Lysis Buffer (R2841)
  • Wash Buffer 1 (conc.)(R2851)
  • Wash Buffer 2 (conc.)(R2861)
  • DNase l (lyophilized)
  • DNase I Reconstitution Buffer
  • 2x DNase I Buffer
  • Manganese Chloride Solution
  • Water, nuclease-free
  • GeneJET RNA Purification Columns pre-assembled with Collection Tubes
  • Collection Tubes, 2 mL
  • Collection Tubes, 1.5 mL
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C-8°C.
Format Spin Column
Isolation Technology Spin Column
Purification Time 30 min.
Sample Type Blood
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Nucleic Acid Labeling, Northern Blotting, cDNA Library Construction
No. of Reactions 50 Preps
Product Line GeneJET
Quantity 50 preps
Target Total RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.