Learn More
Applied Biosystems™ GeneMapper™ PG Software v1.0
Product Code. 31369207 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 License
Product Code. Quantity
31369207 1 License
1 options
Product Code. 31369207 Supplier Applied Biosystems™ Supplier No. A77500
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
GeneMapper PG Software v1.0 is a probabilistic genotyping application that transforms complex DNA mixture interpretation.
GeneMapper PG Software v1.0 is a probabilistic genotyping application that transforms complex DNA mixture interpretation. Using a continuous model running a deterministic mixture deconvolution algorithm, GeneMapper PG Software delivers intuitive, reproducible, and transparent results. It provides real-time explanations and visualizations to drive clarity and understanding during each DNA mixture analysis step.
GeneMapper PG Software v1.0 features
  • Compare profiles—compare profiles across your project
  • Number of contributors (NOC) estimation—perform straightforward analysis (maximum allele count (MAC) and total allele count (TAC)) and more advanced analyses, such as machine learning-based decision trees for accurate estimation
  • Deconvolution—a continuous model using a deterministic algorithm providing precise, reproducible mixture results
  • Likelihood ratio (LR) calculations and robustness analysis—support for simple to complex scenarios, including assumed contributors and multiple potential contributors, along with simulation analysis to evaluate significance of results
GeneMapper PG Software v1.0 offers seamless integration with GeneMapper ID-X Software v1.7 for easy transfer of data across your workflow. Additionally, multiple file types are supported, allowing broad compatibility with a variety of data sources. Compatible file formats include: .ser project files from GeneMapper ID-X Software, CODIS CMF files formats 3.2 and 3.3, and .csv and .txt file types. This versatility allows for efficient and comprehensive data management, enhancing the overall workflow and compatibility with existing systems.
GeneMapper PG Software v1.0 employs a deterministic method that mirrors manual analysis. Complex formulas and calculations are accessible, with the user interface displaying the logic in real-time. Consistency allows the results to remain the same with repeat analysis. Powerful visualization tools help explain results and allow detailed analysis exploration. GeneMapper PG Software can help enhance forensic analysis by providing clear, insightful answers, enabling informed decisions with confidence.
TRUSTED_SUSTAINABILITY
Specifications
Software Type License
Starting Material DNA
Product Type Software License
Quantity 1 License