missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genepaddles™ Electroporation Electrodes
Description
- Compatible with BTX Gemini X2 and ECM-2001 electroporation systems
- Two models to suit the size of the target area
- Gold plated, noninvasive
- Thickness: 1 mm
- Easily cleaned
- Life span is approximately 1500 pulses
- 450122 and 450123 require 45-0203 holder and 45-0216 cables (not included)
Specifications
Specifications
| For Use With (Application) | In-Vivo Gene Delivery, in-Vitro Embryo Gene Delivery |
| For Use With (Equipment) | ECM 830, ECM 2001+, Legacy ECM 2001, and Gemini X2 |
| Material | Gold-plated |
| Product Line | BTX Electroporators |
| Product Type | Genepaddles Kit |
| Voltage | 200 V |
| Electrode Shape | Rectangular |
| Includes | Micrograbber cable, square post cable, banana plug and banana cable, genetrode holder, genepaddle |
| Length (English) | 0.19 in. |
| Length (Metric) | 5 mm |
| Show More |
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?