missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ General Laboratory Tubing

Product Code. 11522583 Shop All Fisherbrand Products
Change view
Click to view available options
Overall Length (Metric):
5 m
Unit Size:
5m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) unitSize
11522583 5 m 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11522583
Only null left
Add to basket
Add to basket

Used for transferring corrosive liquids, even at high temperatures

  • Shows chemical resistance, particularly to aggressive fuels, acids, animal and vegetable oils, solvents, alcohols and bleaching agents
  • Good resistance to ozone and radiation
  • Other sizes are available on request

Specifications

Material Viton
Overall Length (Metric) 5 m
Thickness (Metric) 0.75 mm
For Use With (Application) Transferring corrosive liquids, even at high temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.