missing translation for 'onlineSavingsMsg'
Learn More

Manutan General masking tape, LxW 50m x 38mm

Product Code. 12899139
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12899139 Supplier Manutan Supplier No. 193D041N

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Highly conformable general purpose masking tape that can be removed cleanly and easily. Ideal for spray masking and many other uses from the drawing studio to the factory.
  • Roll length : 50m
  • Width : 38mm
  • Pack quantity : 24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.