Learn More
Thermo Scientific™ General Purpose Centrifuge Rotor Packages
Product Code. 16390778 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
TX-1000 Blood Tube Package
TX-1000 Cell Culture Package
TX-1000 Microplate Package
TX-200 Blood Tube Package
TX-200 Cell Culture Package
TX-400 Blood Tube Package
TX-400 Cell Culture Package
TX-750 Blood Tube Package
TX-750 Cell Culture Package
Unit Size:
1 unit
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
16390778 TX-400 Blood Tube Package 1 unit
16350778 TX-1000 Blood Tube Package 1 unit
16360778 TX-1000 Microplate Package 1 unit
16370778 TX-1000 Cell Culture Package 1 unit
16380778 TX-200 Cell Culture Package 1 unit
16300788 TX-400 Cell Culture Package 1 unit
16310788 TX-750 Blood Tube Package 1 unit
16320788 TX-750 Cell Culture Package 1 unit
16330788 TX-200 Blood Tube Package 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 16390778 Supplier Thermo Scientific™ Supplier No. 75016107
  • ONLINE EXCLUSIVE PRICE
    5385.00 €
    5200.00 € / 1 units
    Save 185.00 €
    3% Off
View all promotions
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Everything you need for your swing out rotor in one package
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
TRUSTED_SUSTAINABILITY
Specifications
Material See individual centrifuge, rotor and accessory data
Type Rotor Equipment Package
Capacity Max in TX-400: 76 x 5/7 mL Blood Tubes, 56 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
Net Weight See individual components of package
For Use With (Equipment) Purchase of Thermo Scientific General Purpose Centrifuge required for operation
Certifications/Compliance EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Includes TX-400 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Warranty See Manual
Biocontainment Yes
Model TX-400 Blood Tube Package