missing translation for 'onlineSavingsMsg'
Learn More

3M™ General Purpose Masking Tape 201E

Product Code. 17843256
Change view
Click to view available options
Dimensions (L x W):
50 m x 24 mm
50 m x 48 mm
Unit Size:
Pack of 24
Pack of 36
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17843256 50 m x 48 mm Pack of 24
17833256 50 m x 24 mm Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17843256 Supplier 3M™ Supplier No. 7100195888
  • ONLINE EXCLUSIVE PRICE
    140.00 €
    111.23 € / Pack of 24
    Save 28.77 €
    20% Off
Estimated Shipment: 04-09-2026
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

The 3M™ General Purpose Masking Tape 201E is a premium-quality masking tape used for short-term paint masking, fixing, bundling, holding, sealing, and other applications up to a temperature of 80°C for one hour.

  • This tape is designed for general use in industrial applications with a slight need for temperature and water resistance such as short-term paint masking and sealing applications.
  • It provides a good instant adhesion and holding power that resists lifting or curling.
  • Its crepe paper backing offers conformability that will maintain integrity when formed around corners.
  • Its good solvent and moisture resistance helps to resist paint bleed through and provides good paint lines due to its clean removability in one piece with no residue from most surfaces.
TRUSTED_SUSTAINABILITY

Spécification

Color Beige
Material Crepe Paper
Product Type Masking Tape
Dimensions (L x W) 50 m x 48 mm
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.