Learn More
3M™ General Purpose Masking Tape 201E
Beskrivelse
- This tape is designed for general use in industrial applications with a slight need for temperature and water resistance such as short-term paint masking and sealing applications.
- It provides a good instant adhesion and holding power that resists lifting or curling.
- Its crepe paper backing offers conformability that will maintain integrity when formed around corners.
- Its good solvent and moisture resistance helps to resist paint bleed through and provides good paint lines due to its clean removability in one piece with no residue from most surfaces.
Tekniske data
Tekniske data
| Color | Beige |
| Material | Crepe Paper |
| Product Type | Masking Tape |
| Dimensions (L x W) | 50 m x 24 mm |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?