missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 120 LIZ™ dye Size Standard

Product Code. 15839766 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
800 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839766 800 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15839766 Supplier Applied Biosystems™ Supplier No. 4324287
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Contains 9 single stranded labeled DNA fragments

The GeneScan™ 120 LIZ™ Size Standard is a five dye-labeled size standard that is designed for reproducible sizing of small fragment analysis data.

The GeneScan™ 120 LIZ™ Size Standard is a five dye-labeled size standard that is designed for reproducible sizing of small fragment analysis data.

GeneScan™ 120 LIZ™ Size Standard is designed for sizing DNA fragments in the 15-120 nucleotides range and provides nine single-stranded labeled fragment of: 15, 20, 25, 35, 50, 62, 80, 110 and 120 nucleotides. The sizing curve generated from these short fragments make the GeneScan™ 120 LIZ™ Size Standard ideal for use with the ABI Prism SNaPshot™ Multiplex Kit as well as other short fragment analysis applications. Each of the DNA fragments is labeled with the LIZ™ fluorophore which results in a single peak when run under denaturing conditions. With the 5th dye LIZ™ your marker fragments can be labeled with the dyes FAM™, VIC™, NED™ or PET™.

Since the standard is labeled with the fifth dye, you can genotype a greater number of markers in a given lane, compared to the four-dye system.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 9 single stranded labeled DNA fragments. Store the kit at 2° to 8°C. (Do not freeze).
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
Product Line GeneScan, LIZ
Type Dye Size Standard
Label or Dye LIZ
No. of Reactions 800 Reactions
Quantity 800 reactions
Shipping Condition Gel Pack

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.