missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 500 LIZ™ dye Size Standard

Product Code. 15843570 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
800 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15843570 800 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15843570 Supplier Applied Biosystems™ Supplier No. 4322682
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Contains 16 LIZ labeled single stranded DNA fragments for use as a standard

The GeneScan™ 500 LIZ™ Size Standard is a fifth dye-labeled size standard for the reproducible sizing of fragment analysis data.

The GeneScan™ 500 LIZ™ Size Standard is a fifth dye-labeled size standard for the reproducible sizing of fragment analysis data.

GeneScan™ 500 LIZ™ Size Standard is designed for sizing DNA fragments in the 35-500 nucleotides range and provides 16 single-stranded labeled fragment of: 35, 50, 75, 100, 139, 150, 160, 200, 250, 300, 340, 350, 400, 450, 490 and 500 nucleotides. The sizing curve generated from these fragments make the GeneScan™ 500 LIZ™ Size Standard ideal for a variety of fragment analysis applications such as Microsatellites, Fragment Length Polymorphisms and Relative Fluorescent Quantitation. Each of the DNA fragments is labeled with the LIZ™ fluorophore which results in a single peak when run under denaturing conditions. With the 5th dye LIZ™ your marker fragments can be labeled with the dyes FAM™, VIC™, NED™ or PET™.

Since the standard is labeled with the fifth dye, users can genotype a greater number of markers in a given lane, compared to the four-dye system.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Compatibility Capillary Electrophoresis
For Use With (Application) Fragment analysis
Quantity 800 reactions
Product Line GeneScan, LIZ
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
Label or Dye LIZ
Content And Storage Contains 16 LIZ labeled single stranded DNA fragments for use as a standard. Store the kit at 2°C to 8°C (Do not freeze).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.