missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 500 ROX™ dye Size Standard

Product Code. 15849746 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
800 Reactions
9,600 injections
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849746 9,600 injections Each
15829716 800 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15849746 Supplier Applied Biosystems™ Supplier No. 4310361
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

16 ROX™ dye-labeled, Single-stranded DNA fragments

The GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems.

The GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems. The use of an internal lane size standard enables automated data analysis and is also essential for achieving high run to run precision in sizing DNA fragments by electrophoresis. GeneScan™ 500 ROX™ Size Standard is designed for sizing DNA fragments in the 35-500 bp range and provides 16 single-stranded ROX™ dye labeled fragments of 35, 50, 75, 100, 139, 150, 160, 200, 250, 300, 340, 350, 400, 450, 490, and 500 bases. Each of the DNA fragments is labeled with a proprietary fluorophore, which results in a single peak when run under both denaturing and non-denaturing conditions.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Sequencing
Quantity 9,600 injections
Product Line GeneScan, ROX
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
Label or Dye ROX
Product Type Dye
Run Time Standard
Content And Storage This kit contains ROX size standard and loading buffer.
Store the kit at 2°C to 8°C. (Do not freeze).
Description GeneScan™ 500 ROX™ dye Size Standard, ROX™ Label. Use with 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.