missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 600 LIZ™ Size Standard v2.0 Dx

Product Code. 15908836 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
800 Reactions
Unit Size:
800 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15908836 800 Reactions 800 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15908836 Supplier Applied Biosystems™ Supplier No. A25794
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this size standard for fragments between 20 and 600 bp. The standard contains 36 LIZ dye-labeled, single-stranded DNA fragments.

The GeneScan 600 LIZ Size Standard v2.0 Dx is a five dye-;labeled, high-density size standard for the reproducible sizing of fragment analysis data. This product can be used as a traditional size standard on all Applied Biosystems capillary electrophoresis systems. Use this size standard for fragments between 20 and 600 bp. The standard contains 36 LIZ dye-labeled, single-stranded DNA fragments. Since the standard is labeled with the fifth dye, users can genotype a greater number of markers in a given lane, compared to the four-dye system.

Size fragments in the 20–600 nucleotides range

GeneScan 600 LIZ Size Standard v2.0 Dx is designed for sizing DNA fragments in the 20–600 nucleotides range and provides 36 single-stranded, labeled fragments of: 20, 40, 60, 80, 100, 114, 120, 140, 160, 180, 200, 214, 220, 240, 250, 260, 280, 300, 314, 320, 340, 360, 380, 400, 414, 420, 440, 460, 480, 500, 514, 520, 540, 560, 580, and 600 bp. The sizing curve generated from fragments make the GeneScan 600 LIZ Size Standard v2.0 Dx ideal for use in a variety of fragment analysis applications such as analysis of microsatellites and fragment length polymorphisms as well as in relative fluorescent quantitation. Each of the DNA fragments is labeled with the LIZ fluorophore, which results in a single peak when run under denaturing conditions. With the fifth dye, LIZ, your marker fragments can be labeled with FAM, VIC, NED, or PET dye.

The GeneScan™ 600 LIZ™ Size Standard v2.0 Dx is a five dye-labeled, high-density size standard for the reproducible sizing of fragment analysis data. This product can be used as a traditional size standard on all of the Applied Biosystems™ capillary electrophoresis systems.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C. Protect from light. Do not freeze.
Format Standard
For Use With (Equipment) 3500 Dx Genetic Analyzer
GC-Rich PCR Performance High
For Use With (Application) Sequencing
Label or Dye LIZ
No. of Reactions 800 reactions
Product Line GeneScan, LIZ
Product Type DNA Sequencing System
Quantity 800 Reactions
Shipping Condition Wet Ice
Size 20 bp, 600 bp
Test Time Fast
Show More Show Less

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.