missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GENESYS G5 Vis and UV-Vis Spectrophotometer Software

Product Code. 32035865 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
BeerCraft Software
Bio Analysis Software
Water Analysis Software
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
32035865 BeerCraft Software Each
32035858 Water Analysis Software Each
32035863 Bio Analysis Software Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32035865 Supplier Thermo Scientific™ Supplier No. 840413800
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ GENESYS™ G5 Software streamlines spectrophotometric workflows with intuitive operation, advanced analysis, and tools for data integrity and compliance.

Thermo Scientific™ GENESYS™ G5 Spectrophotometer Software delivers a flexible, workflow-driven experience that adapts to your laboratory needs, from routine measurements to advanced and regulated environments. With intuitive onboard control, optional PC-based software, and application-specific add-ons, users can streamline analysis and reduce setup time while obtaining consistent results. Data integrity features such as user management, audit trails, and electronic signatures available on-board or via PC support compliance while maintaining efficiency across evolving workflows.

GENESYS G5 software is designed to simplify spectrophotometric workflows while providing flexibility to expand as your lab grows. From intuitive onboard operation to advanced PC control and industry-specific application packages, the platform supports everything from routine measurements to highly regulated environments.

Built for immediate productivity

The onboard software features a modern touchscreen interface that enables users to start measurements quickly with minimal training. Core spectrophotometric capabilities—including absorbance measurements, quantitative analysis, kinetics, scanning, and OD600—are integrated into guided workflows that simplify operation. Methods can be created, saved, and reused, generating consistency across users and experiments.

  • Intuitive touchscreen interface for quick adoption
  • Centralized access to applications and add-ons
  • Customizable, reusable workflows to improve consistency
  • Minimal training required for routine operation

Enables expanded PC control and advanced analysis

Optional PC software extends instrument capabilities beyond the onboard interface, enabling more advanced workflows and enhanced data handling. This allows laboratories to tailor their setup based on complexity, throughput, and reporting needs.

  • Advanced applications including kinetics and quantitation
  • Enhanced reporting and data processing capabilities
  • Flexible control for multi-user or high-throughput labs

Options for application-focused software modules

GENESYS G5 software can be expanded with targeted application packages that simplify complex workflows through pre-configured methods and guided analysis. These modules reduce setup time and improve reproducibility across industries.

  • BeerCraft™ Software – Standardizes brewery QC workflows with built-in ASBC methods for color, bitterness, and consistency
  • Water Analysis Software – Provides pre-configured methods aligned with environmental testing workflows and test kits and option to design custom methods
  • Color Calculation Software – Provides L*a*b*, L*C*h, and other color metrics for fast, repeatable color analysis

Built for flexible data management and connectivity

GENESYS software supports efficient data handling across standalone and connected environments. Users can store, export, and share data through multiple pathways, ensuring accessibility and traceability.

  • Save and export data via USB, network, or Wi-Fi
  • Take advantage of centralized storage for methods, results, and reports
  • Cloud integration
  • Integrate analyses into existing lab data workflows

Designed for regulated environments

Optional built-in compliance tools help laboratories maintain data integrity and meet regulatory requirements as needs evolve. These features are especially valuable in QA/QC and regulated industries.

  • User roles and permissions to control access
  • Audit trails for traceability of actions and changes
  • Electronic signatures for secure data approval
  • Supports compliance with 21 CFR Part 11

Designed to grow with your lab

With modular options, GENESYS G5 software allows labs to start with essential functionality and expand over time. This scalable approach protects investment while enabling adaptation to new applications, workflows, and regulatory demands.

TRUSTED_SUSTAINABILITY

Specifications

Type BeerCraft Software
For Use With (Equipment) GENESYS G5 Vis and UV-Vis Spectrophotometers
Includes Built-in ASBC methods for color, bitterness, and consistency
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.