missing translation for 'onlineSavingsMsg'
Learn More

Geneticin Disulfate Salt, TRC

Product Code. 17974515
Change view
Click to view available options
Quantity:
100 mg
5 g
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974515 100 mg 100mg
17921855 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17974515 Supplier Toronto Research Chemicals Supplier No. TRCG360580100MG
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20 H40 N4 O10 . 2 H2 O4 S
Molecular Weight (g/mol) 692.71
Synonym O-2-Amino-2,7-dideoxy-D-glycero-α-D-gluco-heptopyranosyl-(1→4)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)]-2-deoxy-D-streptamine Sulfate (1:2), Antibiotic G 418 Sulfate, G 418 Sulfate
IUPAC Name (2S,3S,4S,5S)-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(1-hydroxyethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
SMILES CN[C@H]1[C@H](O)[C@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@@]1(C)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H40N4O10.2H2O4S/c1-6(25)14-11(27)10(26)9(23)18(32-14)33-15-7(21)4-8(22)16(12(15)28)34-19-13(29)17(24-3)20(2,30)5-31-19;2*1-5(2,3)4/h6-19,24-30H,4-5,21-23H2,1-3H3;2*(H2,1,2,3,4)/t6?,7-,8+,9+,10+,11-,12-,13-,14+,15+,16-,17-,18+,19-,20+;;/m0../s1
Formula Weight 692.2092
Chemical Name or Material Geneticin Disulfate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.