missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GenEx Professional Software

Product Code. 15990433 Shop All Applied Biosystems Products
Change view
Click to view available options
Content And Storage:
6 Month License Key for 1 User
Includes One 1-year License Key for 1 User
Includes One 2-year License Key for 1 User
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
15990433 Includes One 2-year License Key for 1 User Each
15900443 Includes One 1-year License Key for 1 User Each
15910443 6 Month License Key for 1 User Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15990433 Supplier Applied Biosystems™ Supplier No. A33921
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

GenEx by Multid Analyses is software for the processing and analysis of real-time qPCR data.

Its capabilities range from batch read of data files, basic data pre-processing and management to advanced cutting-edge expression profiling analyses. GenEx software provides methods to select and validate reference genes, classify samples, group genes, monitor time-dependent processes, and much more.

Basic data editing and management
GenEx software v6.1 enables the pre-processing of raw data with consistence, in the correct order, and with confidence. The pre-processing features include interplate calibration, PCR efficiency correction, optimal normalization, genomic DNA background subtraction, outlier detection, and more.

Advanced data analysis
GenEx software offers advanced statistical solutions for post-normalization data analysis. Powerful interactive presentation tools facilitate the generation of professional illustrations of even the most complex experimental designs. Current features include standard curve analyses with estimation of limit of detection (sensitivity), limit of quantification, dynamic range, repeatability and reproducibility, t-test, ANOVA and non-parametric tests with appropriate corrections and estimates of precision, and multivariate statistical tests including Hierarchical Clustering Analysis (HCA), Heatmaps, Principal Component Analysis (PCA), and Artificial Neural Networks (ANN).

GenEx Professional vs. Enterprise
GenEx Professional software provides an extensive set of pre-processing modules together with data visualization and a set of analyses including a larger number statistical analysis modules, suitable for standard curve-based analysis as well as gene expression profiling. GenEx Professional software also contains wizards for direct data import from different instruments.

GenEx Enterprise software contains all modules of GenEx Professional plus the most advanced data analyses, including Dynamic PCA, ROC curve, ANN, and Support Vector Machine (SVM). GenEx Enterprise software also provides modules for data analysis conforming to the Clinical and Laboratory Standards Institute (CLSI) standard documents.

For customers who would like to use GenEx software on multiple PCs, one at a time, GenEx licenses can be pre-installed on a USB key. Install the software on several computers and connect the USB key to the one you want to use at the time. A valid license for GenEx Professional or GenEx Enterprise software is required for purchase of a USB key (Cat. No. A33924).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Includes One 2-year License Key for 1 User
Software Type Real Time PCR Data Analysis
Shipping Condition Room Temperature
Quantity 1 2-year license

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.