missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Genistein

Product Code. 18712804
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18712804 50 mg 50mg
18702704 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18712804 Supplier R&D Systems™ Supplier No. 1110/50
  • ONLINE EXCLUSIVE PRICE
    279.00 €
    263.55 € / 50mg
    Save 15.45 €
    5% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. EGFR kinase inhibitor. Also estrogen and PPARγ ligand

Genistein is a phytoestrogen with a wide range of biological actions. Genistein inhibits protein tyrosine kinases including epidermal growth factor receptor kinase, binds to PPARγ and estrogen receptors and acts as an agonist at GPER. Genistein inhibits aggregation and fibrilization of Aβ42 and amylin37 (hiAPP). In Huntington's disease fibroblast cells, Genistein reduces intracellular mutant huntingtin (mHTT) aggregates and induces degradation of mHTT; in a cellular model of Huntington's disease, Genistein corrects the mutant phenotype through stimulation of autophagy.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-72-0
Molecular Formula C15H10O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 270.24
Purity >98%
Molecular Weight (g/mol) 270.24
Chemical Name or Material Genistein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.