missing translation for 'onlineSavingsMsg'
Learn More

Genistein, TRC

Product Code. 30406879
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406879 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406879 Supplier Toronto Research Chemicals Supplier No. TRCG350000100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Genistein
CAS 446-72-0
Purity Grade Notes HPLC
Molecular Formula C15 H10 O5
Quantity 100 mg
Synonym 4H-1-Benzopyran-4-one, 5,7-dihydroxy-3-(4-hydroxyphenyl)-, Genistein (6CI), Isoflavone, 4',5,7-trihydroxy- (8CI), 4',5,7-Trihydroxyisoflavone, 5,7,4'-Trihydroxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, Baichanin A, Bonistein, C.I. 75610, Climagen F, FW 635I-2, GeniVida, Genisteol, Genisterin, Genistin aglycone, NPI 031L, NSC 36586, Prunetol, SIPI 807-1, Sophoricol
InChI Formula InChI=1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-7,16-18H
SMILES Oc1ccc(cc1)C2=COc3cc(O)cc(O)c3C2=O
IUPAC Name 5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
Molecular Weight (g/mol) 270.24
Formula Weight 270.0528
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.