missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lysis Buffer PL1

Product Code. 11972282
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
This item is not returnable. View return policy

Product Code. 11972282

Brand: Macherey-Nagel™ 740918

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lysis Buffer PL1 to be used with NucleoSpin Plant II kits.

Includes two optimised, alternative lysis buffers based on the established CTAB and SDS lysis methods. As plants are very heterogenous and contain many different metabolites such as polyphenols, polysaccharides, or acidic components, NucleoSpin™ Plant II offers two different lysis proceudres for optimal processing of various samples. The silica membrane in the NucleoSpin™ Plant II columns is optimised to improve DNA binding. NucleoSpin™ filters ar included for convenient clarification of lysates. NucleoSpin™ Plant II allows processing of up to 100 mg (wet weight) or 20mg (dry weight) starting material. Depending on the individual sample typical yields are in the range from 1μg to 30μg DNA. The eluted DNA is ready to use for subsequent reactions such as PCR*, restriction analysis and others. Kit components: NucleoSpin™ Plant II columns, filters, collecting tubes 2mL, buffers, RNase A.
  • Silica membrane technology
  • Yield: 1μg to 30μg
  • Sample size: ≤100 mg wet weight ≤20mg dry weight
  • Elution volume: 100μL
  • Binding capacity: 50μg
  • Preparation time: 30 min./prep
  • NucleoSpin filters included for clarification of lysates
  • RNase A included
  • Format: mini spin column
  • Two alternative lysis buffers included for processing of various samples (PL1 uses CTAB lysis method, PL2 uses SDS lysis method) For isolation of genomic DNA from plant and fungal material.

Specifications

Chemical Name or Material Lysis Buffer PL1
For Use With (Application) For Genomic DNA Isolation
Quantity 125 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.