missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin sulfate, MedChemExpress

Product Code. 16490748
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490748 5 g 5g
16471986 500 mg 500mg
16432029 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490748 Supplier Medchem Express Supplier No. HYA0276
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gentamicin sulfate, an orally active aminoglycoside antibiotic, inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture. Gentamicin sulfate inhibits DNase I with an IC50 of 0.57 mM.

Gentamicin sulfate, an orally active aminoglycoside antibiotic, inhibits the growth of both gram-positive and gram-negative bacteria and to inhibit several strains of mycoplasma in tissue culture. Gentamicin sulfate inhibits DNase I with an IC50 of 0.57 mM.

Specifications

Chemical Name or Material Gentamicin sulfate
CAS 1405-41-0
Color White
Purity Grade Notes Research
Molecular Formula C(19-21)H(39-43)N5O7.H2SO4
Quantity 5 g
Solubility Information H2O : ≥ 30 mg/mL
SMILES O=S(O)(O)=O.O[C@]1(C)C(NC)[C@@H](O)[C@@H](O[C@H]2C(N)C[C@H](N)[C@@H](O[C@@H]3[C@H](N)CC[C@@H]([R])O3)[C@@H]2O)OC1.NC[*].C[C@H]([*])N.C[C@H]([*])NC.[R=].[or].[or]
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.