missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gentamycin M.I.C.Evaluator Strips (M.I.C.E.™)

Product Code. 11404049
Change view
Click to view available options
Description:
Gentamycin M.I.C.Evaluator Strips (M.I.C.E.™)
Concentration:
256-0.015µg/mL
Unit Size:
Pack of 50
Product Code. Description Concentration unitSize
11404049 Gentamycin M.I.C.Evaluator Strips (M.I.C.E.™) 256-0.015µg/mL Pack of 50
1 options
This item is not returnable. View return policy

Product Code. 11404049

Brand: Thermo Scientific™ MA0116F

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Determine accurate Minimum Inhibitory Concentration (MIC) values of antibiotics using MIC testing strips.

Effortlessly establish accurate Minimum Inhibitory Concentration (MIC) values manually using Thermo Scientific™ Oxoid™ Gentamycin M.I.C.Evaluator Strips (M.I.C.E.™).

Conveniently packaged M.I.C.E. strips combine the ease of a diffusion test with the accuracy of an MIC test.

Simple to Perform
  • Distinctive gradient format provides excellent contrast with the agar
  • Increased font size makes reading easier
  • Scale conforms to international standards
  • Strips are tailored to meet clinical requirements while maintaining the accuracy required for research

Easy-to-handle
  • The strip is conveniently presented when the sachet is peeled apart
  • Single-strip packaging eliminates the requirement to maintain and quality control opened strips; open one and use it
  • Stability of the individual M.I.C.E. strips is ensured through the inclusion of a desiccant within each sachet
  • 10 and 50 pack sizes offer users maximum flexibility to put together custom panels for testing
  • Easily stackable boxes simplify storage; all antimicrobials are stored in the fridge (2-8°C)

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 256-0.015µg/mL
Content And Storage 2°C to 8°C
Description Gentamycin M.I.C.Evaluator Strips (M.I.C.E.™)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.